Feather® Stainless Steel Scalpel Blades
Supplier: WORLD PRECISION INSTRUMENTS LLC
504176 504177 504169 504170 504171 504172 504173 504174 504175
77663-536BX 180.06 CAD
77663-536 77663-538 77663-522 77663-524 77663-526 77663-528 77663-530 77663-532 77663-534
Feather® Stainless Steel Scalpel Blades
Scalpels and Scalpel Assemblies Scalpel Blades
Scalpels are used for removing tissue and making initial incisions. Typically, a scalpel is comprised of a blade and a handle. Scalpels are purchase based on the blade number which indicates its shape. The most popular blades are the #10, #11 and #15.
  • Stainless Steel
  • Sterile
The traditional #10 blad has a broad, angled cutting edge. Use this blade for making wide tissue incissions.
The #11 blade is sharp and pointed and used for stabbing or making short incisions. The sharp, pointed tip makes precise shallow cuts. The blade is triangular in shapeMore Product Information