Micro Fine Mesh Baskets with Lids
Supplier: WORLD PRECISION INSTRUMENTS LLC
WP-4348MM WP-4349MM WP-4350MM WP-4351MM WP-4352MM WP-4353MM WP-4354MM WP-4355MM WP-4343MM WP-4344MM WP-4345MM WP-4346MM WP-4347MM
77665-798EA 93.22 CAD
77665-798 77665-802 77665-806 77665-810 77665-812 77665-816 77665-820 77665-824 77665-780 77665-784 77665-788 77665-792 77665-794
Micro Fine Mesh Baskets with Lids
Baskets Wire Baskets
Micro fine baskets are used for washing, sterilizing and storing delicate micro surgical instruments and parts.
  • Manufactured with stainless steel 304
  • Electrolyte polished
  • Top and lid frame wire diameter: 3.5 mm
  • Micro fine mesh with opening squares of 1.0 mm and wire diameter of 0.5 mm
  • Wire locking clip