Reusable Biopsy Punch Kits
Supplier: WORLD PRECISION INSTRUMENTS LLC
504531 504528 504529 504530
77663-676EA 26.8 CAD
77663-676 77663-670 77663-672 77663-674
Reusable Biopsy Punch Kits
Biopsy Punches
The reusable biopsy punches are multi-purpose sampling tools designed with a retractable cutting cannula. They are easy to operate, efficient, safe, fast and precise. Our punches consists of a razor sharp stainless steel cutting tip.
  • Sterilized using ethylene oxide
  • Limited reuse
  • Tip material: 304 stainless steel
It is designed to cut, retrieve and store the cored samples from a source materials. The source could be skin, tissue, gels, paper, cloth, leaves, paint chips, films or other thin, soft substrates.
The biopsy punches are recommended as aMore Product Information