Metzenbaum Scissors, SuperCut
Supplier: WORLD PRECISION INSTRUMENTS LLC
14214 14214-G 14215 14215-G 503269 503270
77661-836EA 211.69 CAD
77661-836 77661-838 77661-840 77661-842 77663-230 77663-232
Metzenbaum Scissors, SuperCut
Scissors
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue.
  • One edge micro serrated, one edge honed to the sharpness of a knife
  • Blunt, narrow tips
  • Stainless steel
The tips are semi-blunt, and they are available in curved or straight styles.