MicroFil™ Flexible Plastic Needles
Supplier: WORLD PRECISION INSTRUMENTS LLC
CMF20G CMF22G CMF23G CMF26G CMF28G CMF31G CMF34G CMF35G CMF90U
77664-128PK 333.38 CAD
77664-128 77664-130 77664-132 77664-134 77664-136 77664-138 77664-140 77664-142 77664-144
MicroFil™ Flexible Plastic Needles
Pipettes Pipette Controllers
MicroFil flexible plastic needles fills micropipettes easily and reliably. Its long and fine tip allows you to start the filling very close to the pipette tip, eliminating both air bubble formation and clogging due to the washing down of dust particles. The transparent amber MicroFil needle is constructed from a combination of plastic and fused silica - no metal components are used. The MicroFil needle can be stored for days with the filling solution inside without clogging.
The MicroFil's tip elasticity is sturdy and very flexible though not unbreakable. Since it is more flexible thanMore Product Information