SafeSeal Reaction Tubes
Supplier: SARSTEDT INC
72.695.001 72.695.002 72.695.004 72.695.005 72.695.006 72.695.007
77588-832CS 282.27 CAD
77588-832 77588-834 77588-836 77588-838 77588-840 77588-842
SafeSeal Reaction Tubes
Tubes PCR Tubes
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • SafeSeal polypropylene caps attached
  • Polypropylene tube material
  • 3 purity grades to meet all requirements
  • Secure seal even under thermal and mechanical stress
  • High centrifugation up to 25,000 ×g
This broad product portfolio in the field of test and micro tubes includes options with round, conical or flat base, with or without skirted base, with push cap or screw cap, with or without print or labels.