Push Caps
Supplier: SARSTEDT INC
65.1245.006 65.809.021 65.809.301 65.809.302 65.809.303 65.809.504 65.1245.002 65.1245.004 65.818 65.717 65.803.305 65.803.312 65.803.332 65.804.001 65.804.300 65.806.001 65.806.306 65.806.307 65.793.314 65.793.317 65.793.318 65.793.320 65.793.516 65.793.519 65.803.003 65.806.308 65.806.310 65.806.311 65.806.312 65.806.513 65.806.611 65.806.999 65.809.001 65.809.005 65.782
77588-042CS 799.31 CAD
77588-042 77588-322 77588-326 77588-328 77588-330 77588-340 77588-036 77588-040 77588-346 77588-218 77588-284 77588-286 77588-288 77588-290 77588-292 77588-294 77588-298 77588-300 77588-262 77588-266 77588-268 77588-270 77588-272 77588-274 77588-278 77588-302 77588-304 77588-306 77588-308 77588-312 77588-314 77588-316 77588-318 77588-320 77588-252
Push Caps
Closures Test/Sample Tube Closures
High-quality, safe, and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • LDPE material
  • Large grip for optimal handling
  • Quick opening and closing of tubes
  • Ribbed surface for maximum grip
Push caps offer excellent sealing properties, and are available in a wide variety of colors to facilitate organization and coding.