SPORTROL® Spore Strips
Supplier: Crosstex
BS-105 PM-106 NMSTS-062
19872-006PK 474.55 CAD
19872-006 19872-008 19872-016
SPORTROL® Spore Strips
Sterilisation Indicators
Biological sterilization indicators are used to perform sterilization cycle validation.
When stored under labeled conditions, all spore strips listed have a shelf life of 18 months, except mini spore strips 19872-014, which have a shelf life of 24 months.
They are created by inoculating spore suspensions of certified population and resistance onto paper strips. Standard spore strips and combination spore strips are 7x38 mm. Mini spore strips are 2x10 mm in size and will fit into small areas of a device where a standard-sized spore strip cannot be used, such as within a syringe barrel, inside tubing, or under a cap.
Certifications: Meet USP and ISO 11138-1 standards.
Ordering information: Spore strips are individually packaged in glassine envelopes in packs of 100. Each shipment includes instructions for use, certified population, D-value, Z-value, survival/kill data, and expiration date.