Reaction Tubes with Soft Push Cap

Supplier: SARSTEDT INC
72.688 72.688.001 72.688.003 72.688.004 72.688.005
77588-708CS 1210.26 CAD
77588-708 77588-710 77588-714 77588-716 77588-718
Reaction Tubes with Soft Push Cap
Tubes PCR Tubes
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.

  • LDPE retaining loop caps attached
  • Polypropylene tube material
  • Smooth, soft stoppers
  • Particularly suitable for frequent opening and closing
  • Color coding ensures better identification
  • Larger packaging units for economical storage and stocking
  • High centrifugation up to 20000×g

This broad product portfolio in the field of test and micro tubes includes options with round, conical or flat base; with or without skirted base; with push cap or screw cap; with or without print or labels.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR