Freezing Point Apparatus, Koehler
Supplier: Koehler
K29700
52932-022EA 3618.63 CAD
52932-022
Freezing Point Apparatus, Koehler
Melting Point Apparatus
Cools aviation gasoline and aircraft turbine fuel samples to determine the temperature at which hydrocarbon crystals appear
Conforms to ASTM D2386 specifications.
Consists of unsilvered vacuum flask, jacketed sample tube, brass packing gland, stirrer, stopper, clamps and stand. Uses acetone or alcohol cooled by solid carbon dioxide as a cooling medium.
Ordering information: Complete with line cord and switch. Mechanical stirrer motor not included.