Bare Platinum Wires
Supplier: WORLD PRECISION INSTRUMENTS LLC
PTP101 PTP201 PTP401 PTP406
77665-030EA 316 CAD
77665-030 77665-032 77665-034 77665-036
Bare Platinum Wires
Wire
Uncoated pure platinum wire is 99.95% pure.
  • Bare platinum wire
  • Available in precut lengths