Multi-Fit Caps for S-Monovette® and Tubes
Supplier: SARSTEDT INC
65.1379.007 65.1379.008 65.1379.010 65.1379
77588-062CS 789.62 CAD
77588-062 77588-064 77588-066 77588-052
Multi-Fit Caps for S-Monovette® and Tubes
Closures Test/Sample Tube Closures
High-quality, safe, and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
  • LDPE material
  • Flexible design can be used for Ø 12 to 17 mm and for S-Monovette® Ø 13mm tubes
  • Different cap colors for better coding
Multi-fit caps offer excellent sealing properties, and are available in a wide variety of colors to facilitate organization and coding.