Liquid Waveguide Capillary Cells, LWCC-M
Supplier: WORLD PRECISION INSTRUMENTS LLC
LWCC-M-10 LWCC-M-100 LWCC-M-50
77664-554EA 4108 CAD
77664-554 77664-556 77664-558
Liquid Waveguide Capillary Cells, LWCC-M
Flow Cells
The LWCC-M series is a MicroLWCC fiber optic low volume flow cell for UV/VIS/NIR absorbance analysis. Based on WPI's established liquid core waveguide technology, the analyte solution functions as the core of a fluid filled light waveguide. Wetted parts in the sample cell light path are PEEK, fused silica and PTFE. Optical fibers are used to transport light to and from the sample cell.
  • High efficiency coupling
  • Low refractive index offset
  • Fits TIDAS systems and other fiber-coupled spectroscopy equipment
  • Fits 500 and 600 µm fibers
  • Wavelength ranges from 200 to 1000 nm
  • DNA, RNA and protein quantification
The cell can be used in biochemistry for DNA, RNA and protein quantification, colorimetric nutrient and trace metal analysis, drug discovery and dissolution testing, process control, and HPLC analysis. PEEK, fused silica, PTFE wetted materials. 1000 psi maximum pressure.