RackSystem for PCR Containers
Supplier: SARSTEDT INC
95.987.002 95.988 95.988.001 95.988.002 95.988.003 95.988.004
77590-008CS 792.86 CAD
77590-008 77590-010 77590-012 77590-014 77590-016 77590-018
RackSystem for PCR Containers
Racks Tube Racks
SARSTEDT PCR consumables are compatible with standard thermocyclers.
  • 96-well format (8×12 spaces) for the use of PCR plates, strips, and individual tubes
  • All individual tubes can be fully transferred into the cycler with the tray of the rack system
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable