ELISA Plates, 96-Well
Supplier: SARSTEDT INC
82.1582.200 82.1583.100 82.1581.100 82.1581.120 82.1581.220 82.1582.100
77589-202CS 359.21 CAD
77589-202 77589-208 77589-184 77589-188 77589-194 77589-200
ELISA Plates, 96-Well
Microplates Cell Culture Plates
ELISA plates are available with two different protein-binding capacities; medium binding and high binding. They are produced in the ANSI/SLAS standard format, making the plates compatible with common dispensers, washers, and readers.
  • Free from DNA/DNase/RNase, free from pyrogens and endotoxins, non-cytotoxic
  • Made of ultra-pure polystyrene in transparent, white, or black for a wide range of applications
  • Transparent ELISA plates are available with three bottom shapes
  • ELISA plates are supplied without lid
ELISA plates are suitable for a wide range of applications in immunoanalytics. Medium binding ELISA plates are hydrophobic, and are generally suitable for adsorption of primarily hydrophobic and larger/flexible molecules. High binding ELISA plates feature a defined hydrophilic surface optimized for the adsorption of more hydrophilic and small/rigid molecules.
Reproducible binding properties due to low variation coefficient (well-to-well, lot-to-lot):
Medium binding: <10%
High binding: <5%