Reaction Tubes with Soft Push Cap

Supplier: SARSTEDT INC
72.689.002 72.689.003 72.689.004 72.689
77588-724CS 1210.26 CAD
77588-724 77588-726 77588-728 77588-720
Reaction Tubes with Soft Push Cap
Tubes PCR Tubes
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world's largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.

  • LDPE retaining loop caps attached
  • Polypropylene tube material
  • Smooth, soft stoppers
  • Particularly suitable for frequent opening and closing
  • Color coding ensures better identification
  • Larger packaging units for economical storage and stocking
  • High centrifugation up to 20,000×g

This broad product portfolio in the field of test and micro tubes includes options with round, conical or flat base; with or without skirted base; with push cap or screw cap; with or without print or labels.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR