Operating Scissors, Curved
Supplier: WORLD PRECISION INSTRUMENTS LLC
501755 501756 501757 501757-G
77662-808EA 90.03 CAD
77662-808 77662-810 77662-812 77662-814
Operating Scissors, Curved
Scissors
Ring-handled operating scissors are a pair of curved surgical scissors commonly used in general surgery for cutting and dissecting tissues.
  • Sharp/sharp, Sharp/blunt, or Blunt/blunt tips
  • Stainless steel or premium German steel
  • Autoclavable
The 11.5 cm length is a smaller size that can be used for a variety of procedures, including biological and medical research. These operating scissors are curved giving a better view of the subject. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries.