Bone Cutters, 4.13"
Supplier: WORLD PRECISION INSTRUMENTS LLC
503252 503253 503254 503255
77663-196EA 92.46 CAD
77663-196 77663-198 77663-200 77663-202
Bone Cutters, 4.13"
Bone Cutters and Rongeurs
Bone cutters have short, thick scissor-style blades, which are designed to efficiently cut through bone tissue.
  • Strong, heavy, cutting tips
  • Choose curved or straight tips
  • Choose serrated or smooth blades
  • Surgical grade stainless steel
Unlike typical scissor blades, these forceps have a more robust, heavier structure to enable effective cutting and shaping of bone during surgical procedures. The ring handles offer a comfortable grip for the surgeon and provide precise control during bone-cutting procedures.
These are 10.5 cm (4.13") long and are available with curved or straight blades, and smooth or serrated tips. The are made of surgical grade stainless steel, which is suitable for chemical sterilization or autoclaving. Clean your surgical instruments promptly after use to ensure a long life.