Mouse/Rat Restrainers
Supplier: WORLD PRECISION INSTRUMENTS LLC
II-78 II-79 II-80 II-81 II-82 II-83 II-84 II-84XL
77664-420EA 1074.4 CAD
77664-420 77664-422 77664-424 77664-426 77664-428 77664-430 77664-432 77664-434
Mouse/Rat Restrainers
Animal Supplies Animal Restraints
Mouse/rat restrainer for use with the non-invasive blood pressure system.
  • Constructed of sturdy clear, ¹/₄" wall acrylic
  • Requires only mild soap and water solution to clean the restrainers (no ammonia-based products should be used)
  • Includes adjustable head gate and the end plate
  • Head gate and end plates can be removed easily to allows the animal to 'walk-in' to the restrainer and avoids any trauma to the specimen
Mouse and rat tail cuff blood pressure measuring systems for preclinicial research, a true turn key system when it is key critical for determining accurate, consistent blood pressure on mice, rats or any other laboratory animal test subject.
A compact, simple yet versatile system that can test from a single up to 200 animals at a time with independent control of each channel. Simple daisy-chaining of systems allow expansion of systems, up to 200 independently, controlled channels. No other system in the world has such vast capabilities as the IITC blood pressure systems. Single animal systems are controlled from the touch screen which allows keying in all necessary test setups. These rodent restrainers are used with the non-invasive blood pressure tail cuff system for mice and rats.
They can also be use for tail vein injections or other studies where the animal needs to be restrained. Eight different sizes are available for use with neonate rats and adult mice up to 1,000 gram rats. Constructed from clear 0.25" wall acrylic, these restrainers are clear and durable. They can be cleaned easily with mild soap and water. The animals are free to walk in, as opposed to traditional methods of bootstrap holders where animals are physically forced into the restrainers backwards, which can cause severe trauma.
Delivery information: All rodent restrainers are supplied with adjustable head gates, and the end plate snaps in place for easy removal. Head gates, end plates and snaps may be purchased separately.
Caution: Do not use ammonia-based solvents for cleaning.