NanoFil™ Needles
Supplier: WORLD PRECISION INSTRUMENTS LLC
NF34BV NF35BL NF35BV NF36BL NF36BV NF33-36BL NF33-36BV NF33BL NF33BV NF34BL
77664-916EA 99.54 CAD
77664-916 77664-918 77664-920 77664-922 77664-924 77664-906 77664-908 77664-910 77664-912 77664-914
NanoFil™ Needles
Syringe Needles
NanoFil™ needles are made for use with WPI's NanoFil™ syringes, are universal to any volume, and are available in both blunt and beveled styles.
  • Tip material: Stainless steel
  • Assortment packages are available
Each needle is handcrafted in-house and inspected for quality and consistency for your application. Each needle is fixed to a 30 G base for enhanced rigidity, and allows for a universal fit across any NanoFil syringe. Unique tri-beveled needle style boasts superior sharpness for insertion avoiding mechanical damage of sensitive tissues.
Paired with WPI's gastight NanoFil syringes to see true difference in the application outcomes.