Peri-Star™ Pro Digital Peristaltic Pumps
Supplier: WORLD PRECISION INSTRUMENTS LLC
PERIPRO-2HS PERIPRO-4HS PERIPRO-4LS PERIPRO-8LS
77664-984EA 3570.8 CAD
77664-984 77664-986 77664-988 77664-990
Peri-Star™ Pro Digital Peristaltic Pumps
Pumps Peristaltic Pumps
Peri-Star Pro peristaltic pumps provide accurate and precise pumping with convenience and versatility. This can be run in either flow rate mode (ml/min) or rotation speed mode (rpm). For good laboratory practice, pumps must be calibrated after changing the tubing and solution.
  • Large backlit digital LCD display with water resistant cover
  • Programmable for all tubing sizes between 0.8 and 6.4 mm int.Ø
  • Easy and fast tubing replacement using snap-on cartridges
  • Membrane keypad allows easy programming while protecting controls from fluid entry
  • Actively driven rollers by planetary gears for long lasting tubing life
  • Easy setup and calibration with simple instructions on screen
  • Both high and low flow pumps employ planetary gears for minimal pulsations and greater accuracy
Easily calibrate Peri-Star Pro to deliver flow as accurate as 0.5% in a wide flow range from 0.1 to 280 ml/min. Under rotation speed mode, the digitally controlled stepping motor provides accurate and reproducible operation with 0.1% rpm both forward and in reverse.
Large backlit digital LCD display provides readouts of rotation direction, flow rate or rotation speed, tubing ID, drive status and remote control mode simultaneously. Water resistant membrane keypad allows easy programming while protecting LCD display and controls from fluid entry.
Built-in Human Machine Interface (HMI) with screen instructions in plain English steps you through initial setup, calibration and operating procedures. The user-friendly interface reduces the need to frequently check the printed manual for instruction and reference.
Peri-Star Pro is available in two versions: a 4-roller version for high flow and an 8-roller version for lower volumes while providing high pressure with minimal pulsations.
A unique planetary gear design with eight actively driven rollers (four rollers for higher flow rate model), together with independent tubing compression fine adjustment, enables the high flow accuracy and prolongs tubing life. Snap-on cartridges allow tubing to be changed quickly without cross contamination of solutions.