Microelectrode Holders
Supplier: WORLD PRECISION INSTRUMENTS LLC
MPH6P10 MPH6P12 MPH6P15
77664-862EA 146.94 CAD
77664-862 77664-864 77664-866
Microelectrode Holders
Electrode Holders
WPI's microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers. A Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets. The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps). Filling WPI microelectrode holders with electrolytes containing chloride results in stable electrode potential. Suitable electrolytes include KCl, NaCl and CaCl₂. Holders areMore Product Information