Halsted Mosquito Hemostatic Forceps
Supplier: WORLD PRECISION INSTRUMENTS LLC
15920 15920-G 15921 501291 501291-G 501705 501705-G
77661-948EA 92.46 CAD
77661-948 77661-950 77661-952 77662-566 77662-568 77662-672 77662-674
Halsted Mosquito Hemostatic Forceps
Forceps
Halsted (or Halstead) Mosquito Forceps, also called mosquito hemostatic forceps or simply hemostats, are surgical instruments designed for clamping blood vessels or other small tissues to control bleeding during surgery or to provide a secure grip. They are so named because they resemble the slender legs of a mosquito.
  • Halsted Mosquito Forceps are smaller than standard mosquito forceps
  • These are available in 12.5 cm (5") or 9 cm (3.5") lengths
  • Serrated, fine tips
Life science researchers like these fine hemostats because of the secure grip and theMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable