Chemical Resistant Extended Sleeve Gloves, 719
Supplier: Showa
719-07 719-08 719-09 719-10 719-11
77623-456CS 406.09 CAD
77623-456 77623-458 77623-460 77623-462 77623-464
Chemical Resistant Extended Sleeve Gloves, 719
Gloves
Protecting against a broad range of hazards, the SHOWA 719 chemical-resistant gloves feature an extended gauntlet for forearm protection.
  • 100% Nitrile, ergonomic, unsupported and unlined
  • Chemical, water and abrasion resistant
  • Disposable
  • Flexible with forearm protection
  • Oil-resistant
  • The diamond grip makes wet work easier and safer
The 0.28 mm thick unsupported nitrile provides resistance against chemicals, solvents, oils, and animal fats. The ergonomic design ensures comfort during long periods of wear.
Applications: Light chemical handling in horticulture and agriculture, food packing and handling poultry, meat and seafood processing washing and cleaning laboratory, pharma and analysis fruit and vegetable processing.
Certifications: CE, Cat III, EN 388:2016+A1:2018, EN ISO 374- 1:2016+A1:2018/Type, EN ISO 374-5:2016 and Food Contact.
Packaging: 12 Pair per polybag, 12 Polybags per case, 144 Pair per case.