Operating Scissors, Disposable
Supplier: WORLD PRECISION INSTRUMENTS LLC
503713-12 503727-12 503714-12 503715-12 503716-12 503717-12 503718-12
77663-374PK 64.78 CAD
77663-374 77663-400 77663-376 77663-378 77663-380 77663-382 77663-384
Operating Scissors, Disposable
Scissors
Ring-handled operating scissors are surgical scissors commonly used in general surgery for cutting and dissecting tissues. Versatile size that can be used for a variety of procedures, including biological and medical research. These operating scissors are available with straight or curved blades. Our operating scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. These surgical scissors are available with a variety of tips, including two sharp pointed blades, one sharp and one blunt blade, or two blunt blades.
  • Great for one-time use or students
  • Straight or curved blades
  • Sharp/sharp, sharp/blunt, or blunt/blunt tips
Our operating scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. These economy scissors are sold in packages of 12, perfect for students or for disposable use. All sales are final on disposable instrument packs.