Mayo Scissors Supercut
Supplier: WORLD PRECISION INSTRUMENTS LLC
14216 14216-G 14217-G 503384 503278 503279 503280 503281 503277
77661-844EA 211.69 CAD
77661-844 77661-846 77661-848 77663-310 77663-236 77663-238 77663-240 77663-242 77663-234
Mayo Scissors Supercut
Scissors
Supercut mayo scissors, also called mayo dissecting scissors, are surgical scissors made popular by the mayo clinic. They have ring handles and long, straight, thick blades with blunt tips that are straight or curved. The surgical scissors vary in length from 14 cm (5.5") to 25 cm (9.8"). Longer blades give you greater reach for deeper dissection. The blades are sturdy for heavy duty work like cutting connective tissues, fascia, and sutures. Mayo scissors are often used for dissection, where the blunt tips prevent accidental damage to surrounding tissue and may be used to delicately probe.More Product Information