SHOWA CS700 Chemical Resistant Gloves
Supplier: Showa
CS700L-09 CS700M-08 CS700S-07 CS700XL-10 CS700XXL-11
CA76521-122CS 790.48 CAD
CA76521-122 CA76521-124 CA76521-126 CA76521-128 CA76521-130 76521-126 76521-124 76521-122 76521-128 76521-130
SHOWA CS700 Chemical Resistant Gloves
Gloves
With anti-slip technology, the CS700 food-safe gloves offer ice-grip and increased tactility. A double-dipped nitrile coating provides chemical protection and ensures the gloves are highly durable to keep the wearer safe from harmful substances. The combination of a seamless liner and natural latex-free design is kind to the skin.
  • Anti-slip grip
  • Chemical and oil resistant
  • Seamless knit
  • Robust durability
  • Natural rubber latex-free
  • Double coated Nitrile
  • Polyester liner
  • Rough grip
Trades and Applications:
Chemical spray and treatment
Light Chemical Handling in Horticulture, Agriculture
Coating preparation
Laboratory, pharma and analysis
Painting and spray workshops
Fruit and vegetable processing
Grain, mill and starch products
Washing and cleaning
Certifications: Cat III 0598, EN 388:2016+A1:2018 3132A, EN ISO 374-1:2016+A1:2018/Type A JKLMNOPT, EN ISO 374-5:2016, EN 407:2004 X1XXXX, Food Contact EN ISO 21420:2020.
Packaging: Pair per polybag: 12, Polybags per case: 6, Pair per case: 72.