Standard Fluorometer Cell, 10 mm Path, Style A
Supplier: WORLD PRECISION INSTRUMENTS LLC
CUV2051-1 CUV2052-1
77664-168EA 522.98 CAD
77664-168 77664-170
Standard Fluorometer Cell, 10 mm Path, Style A
Cuvettes Spectrophotometer Cuvettes
Glass and synthetic quartz cuvettes are ideal for UV/VIS/NIR absorbance or fluorescence experiments. Synthetic quartz can be used in deep UV applications and is recommended for fluorescent applications, as it does not exhibit background fluorescence. Quartz cuvettes (absorbance, fluorescence and flow) are shipped individually packaged. These economic quartz are ideal for precision measurements because of their high quality materials used and their low manufacturing tolerances.
Spectral Range (>80%): 200 to 2500 nm; Transmission Difference between different cuvettes: Less than 1% (Typical transmission curves of synthetic quartz cuvettes were measured empty, thickness 1.25 mm × 2, including surface reflections, measured with a TIDAS II against air as reference).