Metzenbaum Scissors, Tungsten Carbide
Supplier: WORLD PRECISION INSTRUMENTS LLC
501733 501733-G 501734 501734-G 501739 501739-G 501740 501744 501744-G 501745 501745-G
77662-748EA 313.88 CAD
77662-748 77662-750 77662-752 77662-754 77662-756 77662-758 77662-760 77662-768 77662-770 77662-772 77662-774
Metzenbaum Scissors, Tungsten Carbide
Scissors
The shaft of metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.
  • Tungsten carbide inserts