Iris Scissors, Supercut
Supplier: WORLD PRECISION INSTRUMENTS LLC
14219-G 14224 14224-G 14225 14225-G 14218 14218-G 14219 503262 503259 503260 503261
77661-856EA 243.3 CAD
77661-856 77661-866 77661-868 77661-870 77661-872 77661-850 77661-852 77661-854 77663-216 77663-210 77663-212 77663-214
Iris Scissors, Supercut
Scissors
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
  • Supercut-one edge micro serrated, one edge honed to the sharpness of a knife
  • Sharp tips