Meyhoefer Curette
Supplier: WORLD PRECISION INSTRUMENTS LLC
501773 504769 504770 504771 504772
77662-838EA 94.91 CAD
77662-838 77663-782 77663-784 77663-786 77663-788
Meyhoefer Curette
Curettes
The Meyhoefer curette has an ice cream scoop-shaped tip with semi-sharp edges designed for scraping. In ophthalmology, it may be used to scrape chalazion from the eyelids. It is perfect for incising cysts, large and small.
  • Flat handle a for solid grip
  • Stainless steel
This curette has a sturdy handle with a smooth surface that improves the grip. Its square shape give you a solid grip and helps alleviate finger strain.