Gel Filtration Calibration Kits, Cytiva
Supplier: Cytiva
28-4038-41
28-4038-42
CA95017-646LEA
1884.79
CAD
CA95017-646L
CA95017-648L
Gel Filtration Calibration Kits, Cytiva
Protein Purification
Protein Purification Kits
Gel Filtration Calibration Kits are suitable for calibration of high-resolution gel filtration media and columns such as Superdex, Superose, and Sephacryl.
- An optimized range of proteins that suits high-resolution media and prepacked columns
- Low and high molecular weight ranges from 6500 to 75000 and 43000 to 669000
- Each kit contains five proteins that are lyophilized in individual vials
- Includes Blue Dextran 2000 to determine the void fraction in the column (also available separately)
Caution: For research use only