Mayo Scissors, Tungsten Carbide
Supplier: WORLD PRECISION INSTRUMENTS LLC
500218-G 500219 500219-G 500220 500220-G 500221 500221-G 500218
77662-176EA 199.08 CAD
77662-176 77662-178 77662-180 77662-182 77662-184 77662-186 77662-188 77662-174
Mayo Scissors, Tungsten Carbide
Scissors
Mayo scissors come various lengths. Their construction is extremely durable, and Mayo scissors are used to cut connective tissues, fascia and sutures, primarily. The tips are semi-blunt, and they are available in curved or straight styles.
  • Tungsten carbide insert
  • Stainless steel