Standard Rectangular Quartz Cuvettes
Supplier: WORLD PRECISION INSTRUMENTS LLC
CUV2011-1 CUV2012-1 CUV2101-1 CUV2102-1 CUV2105-1 CUV2106-1 CUV2108-1
77664-156EA 178.54 CAD
77664-156 77664-158 77664-178 77664-180 77664-182 77664-184 77664-186
Standard Rectangular Quartz Cuvettes
Cuvettes Spectrophotometer Cuvettes
Glass and synthetic quartz cuvettes are ideal for UV/VIS/NIR absorbance or fluorescence experiments. Synthetic quartz can be used in deep UV applications and is recommended for fluorescent applications, as it does not exhibit background fluorescence. Quartz cuvettes (absorbance, fluorescence and flow) are shipped individually packaged. These economic quartz are ideal for precision measurements because of their high quality materials used and their low manufacturing tolerances.
Spectral Range (>80%): 200 to 2500 nm; Transmission Difference between different cuvettes: Less than 1% (More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable