Tenotomy Scissors
Supplier: WORLD PRECISION INSTRUMENTS LLC
14220 14220-G 14221 14221-G 14395-G 14396 14396-G 14395 503263 503264 503265 503266 503267 503268
77661-858EA 231.15 CAD
77661-858 77661-860 77661-862 77661-864 77661-896 77661-898 77661-900 77661-894 77663-218 77663-220 77663-222 77663-224 77663-226 77663-228
Tenotomy Scissors
Scissors
Tenotomy scissors are used for delicate procedures. With the long handle and thin tapered blades, the tenotomy scissors are able to cut tissues in hard to reach, limited spaces.
  • One serrated blade, one honed razor sharp
  • Sharp tips
  • Surgical grade stainless steel
These sharp, straight scissors make small, precision cuts, and they are perfect for delicate operations. The longer handles help to control the precision of these tenotomy scissors in restricted spaces or with delicate membranes or tissues, like brain tissue or ophthalmic surgeries.
These scissors are designed for probing and cutting in tight spots. A precision pair of scissors like the tenotomy scissors limits the damage to surrounding tissues, which is critical in neurosurgery and other applications.