Crimped Wire Mesh Sterilization Baskets, Tilted Handles
Supplier: WORLD PRECISION INSTRUMENTS LLC
WP-4404CW WP-4406CW WP-4427CW WP-4430CW WP-4415CW WP-4424CW WP-4425CW WP-4431CW WP-4436CW WP-4438CW WP-4401CW WP-4402CW WP-4411CW WP-4412CW WP-4413CW WP-4407CW WP-4410CW WP-4418CW WP-4422CW WP-4444CW WP-4445CW
77665-880EA 77.42 CAD
77665-880 77665-890 77666-000 77666-014 77665-942 77665-984 77665-990 77666-020 77666-042 77666-052 77665-864 77665-870 77665-920 77665-926 77665-932 77665-896 77665-912 77665-956 77665-974 77666-078 77666-084
Crimped Wire Mesh Sterilization Baskets, Tilted Handles
Baskets Wire Baskets
Stainless steel crimped wire mesh sterilization baskets with tilt handles.
  • Electropolished
  • Burr free to protect wraps and prevent injury
  • Smooth rounded corners
  • Tilt handles are mounted on the inside and drop inside the basket for stacking
  • Top and bottom frame diameter: 5.0 mm
  • Crimped mesh with mesh openings of 5 mm, wire Ø of 0.9 mm
Stackable baskets meet all requirements for automatic washing and sterilization of all instrument types.
Applications: Instrument cleaning, disinfection and sterilization and instrument storage and handling.