Semi-Micro, Self-Masking Quartz Cuvette, 10 mm, Style D
Supplier: WORLD PRECISION INSTRUMENTS LLC
CUV2031-1 CUV2032-1 CUV2033-1 CUV2034-1
77664-160EA 413.61 CAD
77664-160 77664-162 77664-164 77664-166
Semi-Micro, Self-Masking Quartz Cuvette, 10 mm, Style D
Cuvettes Spectrophotometer Cuvettes
WPI’s glass and synthetic quartz cuvettes are ideal for UV/VIS/NIR absorbance or fluorescence experiments. Synthetic quartz can be used in deep UV applications and is recommended for fluorescent applications, as it does not exhibit background fluorescence. Quartz cuvettes (absorbance, fluorescence and flow) are shipped individually packaged. These economic quartz are ideal for precision measurements because of their high quality materials used and their low manufacturing tolerances.
  • 12.5×12.5×45 mm