Notrax® 765 Deep Freeze Mat
Supplier: NOTRAX USA, INC.
765C0036BL168 765C0036BL240 765C0036BL72 765S0048BL
77661-458EA 1342.4 CAD
77661-458 77661-460 77661-462 77661-464
Notrax® 765 Deep Freeze Mat
Floor Mats Traction and Anti-Fatigue Floor Mats
The NoTrax Deep Freeze Mat™ 765 provides the ultimate solution for keeping your freezer floor clean, safe, and slip-free. This innovative freezer mat is engineered to withstand extreme cold temperatures while providing exceptional performance and durability.
  • Made from a tough rubber compound
  • Raised knob solid top surface that enhances traction
  • Tested to withstand temperatures to −40 °F (approximately −40 °C)
  • Manufactured with Microstop anti-microbial rubber compound that inhibits growth of bacteria and fungi
  • Low profile allows for easy on/off cart traffic
More Product Information