ALM Self-Retaining Retractors
Supplier: WORLD PRECISION INSTRUMENTS LLC
14240 14240-G
77661-878EA 153.31 CAD
77661-878 77661-880
ALM Self-Retaining Retractors
Retractors
The ALM (Andrews-Lucia-McPherson) self-retaining retractor is a surgical instrument designed to hold the eyelids open during ophthalmic surgery, providing surgeons with a clear view of the eye.
  • 7 cm (2.75") long
  • L-shaped prongs 3×3 mm
  • 4×4 prongs
  • Maximum spread of 6 cm
  • Surgical grade stainless steel or premium German steel
The two blades have a curved end that fits into the fornices of the eye. The adjustable blades are held in place by a spring-loaded mechanism. The ALM retractor was designed in the 1960s by three ophthalmologist. It is crafter of surgical grade stainless steel or premium German steel, and they may be autoclaved or sterilized using a chemical sterilant.
Clean your surgical instruments promptly after every use to ensure a long life for your surgical tools.