Connectors, Wire to Mini Banana
Supplier: WORLD PRECISION INSTRUMENTS LLC
2033 2034 2035
77662-026EA 55.98 CAD
77662-026 77662-028 77662-030
Connectors, Wire to Mini Banana
Fittings and Connectors
Banana plugs are often used for making connections with WPI amplifiers. This is a mini, insulated, male banana plug connector. The wire is held in place with an internal set screw. The jack and plug are gold-plated to resist corrosion and provide optimal electrical connectivity.
  • Mini banana plug connector (male)
  • Wire held in place with an internal set screw
  • Insulated
  • Rated for 5 A
Jack and plug are gold plated to resist corrosion and provide optimal electrical connectivity.