Spring Scissors, Stainless Steel, Super Cut
Supplier: WORLD PRECISION INSTRUMENTS LLC
WP22201 WP22202B
77665-628EA 391.84 CAD
77665-628 77665-630
Spring Scissors, Stainless Steel, Super Cut
Scissors
Spring scissors, super cut consists of one serrated blade, 100 mm cutting edge.
  • Designed for neurosurgical, vascular and microsurgical uses
  • May be used with either left or right hand
  • Cuts by shearing, reduces crushing of tissue
  • Length of the scissor tips can help with the depth of the incision