Jewelers #5 Forceps
Supplier: WORLD PRECISION INSTRUMENTS LLC
WP6550 WP6560
77666-356EA 116.92 CAD
77666-356 77666-358
Jewelers #5 Forceps
Forceps
Jewelers #5 forceps are precision surgical instruments designed for delicate tasks, particularly in the field of ophthalmic surgery and microsurgery.
  • Straight or angled tip
  • Titanium
Characterized by their fine tips and slender design, these forceps are well-suited for handling small and delicate tissues, such as those involved in eye surgery. The forceps feature knurled round handles, offering a textured surface for a secure grip. This knurled texture enhances the surgeon's control and precision during intricate procedures.
The flat handles offer ergonomic benefits, providing a comfortable grip, which contributes to ease of use during prolonged surgeries without compromising control. Titanium is lightweight, reducing hand fatigue during extended procedures. Additionally, titanium's strength and durability make the forceps resilient to the demands of delicate surgical tasks. These surgical forceps may be sterilized in an autoclave or with a chemical sterilant.
Titanium forceps are 100% non-corrosive (great for seawater procedures) and non-magnetic (MRI compatible), 40% Lighter than stainless steel (reduces hand fatigue), forcep's tips are coated with tungsten carbide for increased gripping power. Anodized, non-glare blue finish.