VWR® 500 Portable pH Meters
Supplier: VWR International
77619-132 77619-134 77619-136 77619-138 77619-140 77619-142 77619-144 77619-146 77619-148
77619-132EA 709.14 CAD
77619-132 77619-134 77619-136 77619-138 77619-140 77619-142 77619-144 77619-146 77619-148
VWR® 500 Portable pH Meters
Electrochemistry Meters pH and Multi-Parameter Meters
The VWR-PH500 meter provide accurate and reliable measurements of pH, mV and temperature in any environment.
  • High resolution LCD display (3.5")
  • 1 to 2 point calibration
  • Automatic identification of three pH standard buffer solutions (4.01, 7.00 and 10.01 pH)
  • IP65 waterproof
  • Automatic or manual temperature compensation capabilities
  • Auto-read and continuous-read measurement options
  • Ability to save 200 sets of measurement results
  • Rechargeable battery
  • Displays electrode status (electrode slope)
  • Three-year meter warranty
The easy to use VWR-PH500 portable meters provide accurate measurements inside and outside the lab with its rechargeable battery. The large, backlit LCD display allows results to easily be viewed during operation.
Ordering information: Carrying case available for purchase separately (77618-810). For application specific recommendations, please reference 'VWR® Electrode Selection Guide' located in the Related Information section.
Delivery information: All meters include electrode holders (2), meter silicone rubber case, and hand strap.