VWR® 700 Portable pH Meters
Supplier: VWR International
77619-170 VWR-PH700AK 77619-174 77619-176 77619-178 VWR-PH700SL VWR-PH700SS VWR-PH700WW 77619-168
77619-170EA 940.14 CAD
77619-170 77619-172 77619-174 77619-176 77619-178 77619-180 77619-182 77619-184 77619-168
VWR® 700 Portable pH Meters
The VWR-PH700 meter provide accurate and reliable measurements of pH, mV and temperature in any environment.
  • High resolution LCD display (3.5")
  • 1 to 5 point calibration
  • IP 65 waterproof
  • Automatic or manual temperature compensation capabilities
  • Auto-read, timed-read, and continuous-read measurement options
  • Ability to save 200 sets of measurement results
  • Rechargeable battery
  • Automatic electrode diagnosis with pH slope and offset display
The easy to use VWR-PH700 portable meters provide accurate measurements in the field and the laboratory. The large, backlit LCD display allows results to easily be viewed during operation. The included carrying case allows for easy transport of all items to make measurements on the go even easier.
Selectable pH buffer groups including NIST, DIN, GB, USA.
Ordering information: For application specific recommendations, please refer 'VWR® Electrode Selection Guide' located in the Related Information section.
Delivery information: All meters include electrode holders (2), meter silicone rubber case, and hand strap. Three-year meter warranty.