VWR® 550-Series Benchtop Conductivity Meters
Supplier: VWR International
VWR-EC550 VWR-EC550A VWR-EC550B VWR-EC550C
77618-956EA 640.94 CAD
77618-956 77618-958 77618-960 77618-962
VWR® 550-Series Benchtop Conductivity Meters
Electrochemistry Meters Conductivity Meters
The VWR-EC550 meter provide accurate and reliable measurements of conducitivty, TDS (total dissolved solids), and temperature in any environment. 
  • High resolution LCD display (6.0")
  • One-point calibration with standard recognition
  • IP54 protection
  • Automatic or manual temperature compensation capabilities
  • Auto-read and continuous-read measurement options
  • Ability to save 50 data sets
  • Ability to set cell constant value
  •  
The easy to use VWR-EC550 benchtop meters provide accurate conductivity measurements within any laboratory environment. Easily choose a kit to fit your application needs by selecting the required cell constant for you sample conductance range.
Ordering information:
Three-year meter warranty. For application specific recommendations, please reference "VWR® Electrode Selection Guide" located in the related information section.  
Delivery information:
All meters include electrode stand and in-use cover.