Hot Plates, Thermo Scientific
Supplier: Thermo Fisher Scientific
HPA2245MQ HPA2235MQ HPA2230MQ HPA2240MQ
33922-804EA 3633.97 CAD
33922-804 33922-757 33922-859 33922-906
Hot Plates, Thermo Scientific
Heaters Hot Plates
These hot plates feature demand-type thermostat controls that will automatically reach and hold temperature to within ±3 °C (5 °F) at 100 °C (212 °F).
  • Temperature range: 38 to 371 °C (100 to 700 °F)
  • Heavy, cast-aluminum top distributes heat evenly
  • Excellent for big heating jobs or multiple smaller ones
Plates feature stepless temperature selection, minimum overshoot, cycle light, and baffled air cooling. Heating element wires are embedded for protection, but are easy to replace if necessary.
Certifications: UL and C-UL listed.