Desmarres Lid Retractors
Supplier: WORLD PRECISION INSTRUMENTS LLC
WP108200 WP108220 WP108230
77665-584EA 77.89 CAD
77665-584 77665-586 77665-588
Desmarres Lid Retractors
Retractors
DeMarres Lid Retractors are surgical instruments used in ophthalmic surgery to hold back the eyelids and expose the eye for examination or surgical procedures. The DeMarres lid retractor is designed to provide a clear and unobstructed view of the eye, particularly during surgeries involving the anterior segment of the eye.
  • DeMarres retractors for ophthalmology
  • Choice of size, 11, 13 or 15 mm wide blade
  • Square handle
  • Surgical grade stainless steel
The blade of the rectractor is curved to follow the contour of the eye, giving optimal exposure with limited discomfort. The tip of the blade is rounded and blunt to avoid injury to delicate eye tissue.
Choose the size of the retractor you need: 0 (11 mm wide), 1 (13 mm wide), and 2 (15 mm wide). The square handles are lightly textured and polished for a solid grip. These retractors are made of surgical grade stainless steel and are suitable for autoclaving.