Liebermann Speculum, Self-Retaining
Supplier: WORLD PRECISION INSTRUMENTS LLC
WP105673 WPI105675
77665-582EA 120.08 CAD
77665-582 77666-418
Liebermann Speculum, Self-Retaining
Specula
The Leibermann Self-Retaining Wire Speculum is used in ophthalmology when examining the globe of the eye.
These specula can hold the eye open during exams or procedures, giving a clear view of the eye structure. This speculum is 7 cm (2.8") long. Choose between 12 and 15 mm wide wire blades. They can open with a maximum spread of 6 cm. Made of stainless steel, these specula are sterilizable.