Fast-Freeze® Flasks and Adapters, Labconco®
Supplier: Labconco
Fast-Freeze®
7542801 7540900 7542300 7547400 7545800 7547000 7541000 7540400 7543300 7545200 7545400 7542701 7543200 7540901 7544200 7542400 7542901 7540300 7542000 7540601 7540600 7542700 7540800 7542600 7545600 7546000 7542601 7542800 7544400 7542900 7542200 7540200 7543000 7541100 7547600 7544000 7540000 7541200 7541101 7545000 7543400 7547200 7540801
72460-040EA 212.87 CAD
72460-040 75841-040 72460-042 72460-044 75841-044 72460-046 89187-296 75841-042 72460-048 75841-048 75841-046 26675-512 26675-319 89187-294 26675-250 26675-230 26675-374 26675-352 26675-396 26675-355 74920-002 72460-050 74920-004 72460-052 82018-226 75841-052 75841-050 72460-036 72460-038 75841-038 26675-501 10148-032 26675-228 26675-308 26675-261 26675-283 26675-240 26675-385 26675-363 26675-341 26675-320 26675-443 26675-487
Fast-Freeze® Flasks and Adapters, Labconco®
Flasks Flat Bottom Flasks
Fast-Freeze® Flasks for FreeZone® Freeze Dry Systems are designed to be easier to handle, faster to load, and more convenient to use in the laboratory for freeze drying.
  • Compatible with all major brands of freeze drying equipment, including Labconco and Virtis™
Fast-Freeze® Flasks simplify lyophilization procedures with a wide-mouth design that allows easy access to samples
Flat bottom, high-strength borosilicate glass flask withstands high heat and deep vacuum. Uniform glass walls maximize heat transfer while maintaining structural strength. Wide-mouth design provides full access to entire inner diameter of glassware for easy loading and cleaning. Silicone rubber snap-on tops provide reliable high-vacuum seals. Flasks should be filled no more than one-third of the volume so a maximum surface area is achieved and efficient lyophilization is assured.
Flask adapters attach to Fast-Freeze® Flasks for connection to valve ports on drying chambers or manifolds for freeze dry systems. Choose borosilicate glass or stainless steel, straight or 45° angle adapters in 0.63 cm (1/4") and 1.9 cm (3/4") diameters.
Flask Holders help to prevent sample melt back. They allow a small Fast-Freeze Flask Bottom containing the sample to be placed inside a larger Fast-Freeze Flask so that the vacuum surrounding the sample insulates it from the room atmosphere. Stainless steel. Flasks not included.
Ordering information: Each complete flask is supplied with 100 filter papers of 2.5 cm (1") diameter. Adapters sold separately. Flask bottoms, tops and filter paper are also available individually. For other glassware including Ampules, Serum Bottles, and Lyph-Lock® Flasks, contact your Avantor representative.