Disposable Safety Scalpels
Supplier: WORLD PRECISION INSTRUMENTS LLC
501337 501338 501339
77662-620PK 119.26 CAD
77662-620 77662-622 77662-624
Disposable Safety Scalpels
Scalpels and Scalpel Assemblies Scalpels
Scalpels are used for removing tissue and making initial incisions.
  • Disposable
  • Ambidextrous - Activation is on the top for right-handed and left-handed users
Typically, a scalpel is comprised of a blade and a handle, though these disposable scalpels are sold as complete units. Scalpels are purchase based on the blade number which indicates its shape. The most popular blades are the #10, #11 and #15.
The traditional #10 blad has a broad, angled cutting edge. Use this blade for making wide tissue incissions.
The #11 blade is sharp and pointed and used for stabbing or making short incisions. The sharp, pointed tip makes precise shallow cuts. The blade is triangular in shape and has the sharp edge along the hypotenuse.
The #15 blade has a curved cutting edge, though not as large as the #10. It is used for making small incisions.